Product Details

SNP ID
rs9621061
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30659283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAACAAGACCTATTCAAAATGC[A/C]TAGGATACAATCAAAATTACACACA
Phenotype
MIM: 611446
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DUSP18 PubMed Links

Gene Details

Gene
DUSP18
Gene Name
dual specificity phosphatase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304794.1 Intron NP_001291723.1
NM_001304795.1 Intron NP_001291724.1
NM_001304796.1 Intron NP_001291725.1
NM_152511.4 Intron NP_689724.3
XM_005261368.4 Intron XP_005261425.1
XM_006724148.3 Intron XP_006724211.1
XM_011529920.2 Intron XP_011528222.1
XM_011529921.2 Intron XP_011528223.1
XM_017028627.1 Intron XP_016884116.1
XM_017028628.1 Intron XP_016884117.1
Gene
SLC35E4
Gene Name
solute carrier family 35 member E4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001479.3 Intron NP_001001479.1
NM_001318370.1 Intron NP_001305299.1
NM_001318371.1 Intron NP_001305300.1
XM_017028794.1 Intron XP_016884283.1
XM_017028795.1 Intron XP_016884284.1

View Full Product Details