Product Details

SNP ID
rs10736156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102259690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATGTATTGATTTAAAAGTGAG[A/C]ATGGCATCAGGAATATCTTTGAAAT
Phenotype
MIM: 603698
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GBF1 PubMed Links

Gene Details

Gene
GBF1
Gene Name
golgi brefeldin A resistant guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199378.1 Intron NP_001186307.1
NM_001199379.1 Intron NP_001186308.1
NM_004193.2 Intron NP_004184.1
XM_005270261.1 Intron XP_005270318.1
XM_006718047.1 Intron XP_006718110.1
XM_006718048.1 Intron XP_006718111.1
XM_006718049.1 Intron XP_006718112.1
XM_006718050.1 Intron XP_006718113.1
XM_011540312.1 Intron XP_011538614.1
XM_011540313.1 Intron XP_011538615.1
XM_011540314.2 Intron XP_011538616.1
XM_017016861.1 Intron XP_016872350.1
XM_017016862.1 Intron XP_016872351.1
XM_017016863.1 Intron XP_016872352.1
XM_017016864.1 Intron XP_016872353.1

View Full Product Details