Product Details

SNP ID
rs12098769
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100273978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAGAAAAATGTTTAAATAGGGG[C/G]CTTATGTTAGGCACGGTGGTTCATG
Phenotype
MIM: 609768 MIM: 616120
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
BLOC1S2 PubMed Links
Additional Information
For this assay, SNP(s) [rs76969433] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
BLOC1S2
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001342.2 1966 UTR 3 NP_001001342.1
NM_001282436.1 1966 UTR 3 NP_001269365.1
NM_001282437.1 1966 UTR 3 NP_001269366.1
NM_001282438.1 1966 UTR 3 NP_001269367.1
NM_001282439.1 1966 UTR 3 NP_001269368.1
NM_173809.4 1966 UTR 3 NP_776170.2
XM_017016102.1 1966 Intron XP_016871591.1
Gene
CWF19L1
Gene Name
CWF19-like 1, cell cycle control (S. pombe)
There are no transcripts associated with this gene.

View Full Product Details