Product Details
- SNP ID
-
hCV31984750
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:175478587 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAAGCGTTTCCGGTGGCGGCGGA[A/G]GCTGCACTGAGCGGGACCTGCGAGC
- Phenotype
-
MIM: 615569
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFXN1
PubMed Links
Gene Details
- Gene
- SFXN1
- Gene Name
- sideroflexin 1
There are no transcripts associated with this gene.
View Full Product Details