Product Details

SNP ID
rs12654727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151182313 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCCACAGACTATCAGGACTGGA[C/G]GGCCTGCTGAGGTTAGTTACTCTAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC69 PubMed Links

Gene Details

Gene
CCDC69
Gene Name
coiled-coil domain containing 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015621.2 2138 UTR 3 NP_056436.2

View Full Product Details