Product Details

SNP ID
rs13160240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137752792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCAACCCCCACCCAGGTTTTG[A/C]TACAGAATCAGCAAGTTCACTCCCT
Phenotype
MIM: 609409
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPA0 PubMed Links

Gene Details

Gene
HNRNPA0
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006805.3 1559 UTR 3 NP_006796.1

View Full Product Details