Product Details
- SNP ID
-
rs3135949
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:48467811 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAAAACACAGAGAAGCCCAAATG[A/G]ATGGAAAGAGTACAGCCTGATTCCA
- Phenotype
-
MIM: 606605
MIM: 607290
MIM: 606609
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATRIP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114997646] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATRIP
- Gene Name
- ATR interacting protein
There are no transcripts associated with this gene.
- Gene
- SHISA5
- Gene Name
- shisa family member 5
- Gene
- TREX1
- Gene Name
- three prime repair exonuclease 1
View Full Product Details