Product Details

SNP ID
rs11846952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22973497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGAGCCGCTGCATGTGGCAACC[A/G]TGGCAGAGCAAGTGCCCATCCAGAG
Phenotype
MIM: 609066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AJUBA PubMed Links

Gene Details

Gene
AJUBA
Gene Name
ajuba LIM protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289097.1 1939 Intron NP_001276026.1
NM_032876.5 1939 Silent Mutation NP_116265.1
NM_198086.2 1939 Silent Mutation NP_932352.1

View Full Product Details