Product Details

SNP ID
rs12280266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124677362 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTAATTTTTATTTTTGTGATGGG[A/G]AACATGTTAGTATATGATTTAAATG
Phenotype
MIM: 610079 MIM: 608621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIAE PubMed Links

Gene Details

Gene
SIAE
Gene Name
sialic acid acetylesterase
There are no transcripts associated with this gene.

Gene
SPA17
Gene Name
sperm autoantigenic protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017425.3 Intron NP_059121.1
XM_011542870.2 Intron XP_011541172.1

View Full Product Details