Product Details

SNP ID
hCV32036670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30326315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGATACTGAGATTCTGTATTT[C/G]TACTGAGGTCTCAGCTGATGCCAGT
Phenotype
MIM: 612295
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL14EP PubMed Links

Gene Details

Gene
ARL14EP
Gene Name
ADP ribosylation factor like GTPase 14 effector protein
There are no transcripts associated with this gene.

View Full Product Details