Product Details

SNP ID
rs4807928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1448181 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAAACCCCACCACGCGCCCTAC[C/T]CCATGGCTCCACCAGACACCCCCTG
Phenotype
MIM: 612034 MIM: 180535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APC2 PubMed Links

Gene Details

Gene
APC2
Gene Name
APC2, WNT signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005883.2 Intron NP_005874.1
XM_005259475.2 Intron XP_005259532.1
XM_006722607.2 Intron XP_006722670.1
XM_006722608.3 Intron XP_006722671.2
XM_006722609.3 Intron XP_006722672.1
XM_006722610.3 Intron XP_006722673.2
Gene
RPS15
Gene Name
ribosomal protein S15
There are no transcripts associated with this gene.

View Full Product Details