Product Details
- SNP ID
-
rs12475914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:216218526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGAAGCCCGCTAAGAGAACAGAA[A/G]TCAAAAACAGTTGGGTTCTAGATGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PKI55
PubMed Links
Gene Details
- Gene
- PKI55
- Gene Name
- DKFZp434H1419
There are no transcripts associated with this gene.
View Full Product Details