Product Details

SNP ID
rs11692810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222679377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCACAGGTGTTTTGATTATTCCA[A/C]GTCCTTATCAGTTCCATATGCATTT
Phenotype
MIM: 610268
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MOGAT1 PubMed Links

Gene Details

Gene
MOGAT1
Gene Name
monoacylglycerol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058165.2 Intron NP_477513.2

View Full Product Details