Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308248.1 | 6340 | UTR 3 | NP_001295177.1 | ||
NM_005665.5 | 6340 | UTR 3 | NP_005656.4 | ||
XM_017002269.1 | 6340 | Intron | XP_016857758.1 | ||
XM_017002270.1 | 6340 | Intron | XP_016857759.1 | ||
XM_017002271.1 | 6340 | Intron | XP_016857760.1 | ||
XM_017002272.1 | 6340 | UTR 3 | XP_016857761.1 | ||
XM_017002273.1 | 6340 | Intron | XP_016857762.1 | ||
XM_017002274.1 | 6340 | Intron | XP_016857763.1 | ||
XM_017002275.1 | 6340 | Intron | XP_016857764.1 | ||
XM_017002276.1 | 6340 | Intron | XP_016857765.1 | ||
XM_017002277.1 | 6340 | Intron | XP_016857766.1 | ||
XM_017002278.1 | 6340 | Intron | XP_016857767.1 | ||
XM_017002279.1 | 6340 | Intron | XP_016857768.1 | ||
XM_017002280.1 | 6340 | Intron | XP_016857769.1 | ||
XM_017002281.1 | 6340 | Intron | XP_016857770.1 | ||
XM_017002282.1 | 6340 | Intron | XP_016857771.1 | ||
XM_017002283.1 | 6340 | Intron | XP_016857772.1 | ||
XM_017002284.1 | 6340 | Intron | XP_016857773.1 | ||
XM_017002285.1 | 6340 | Intron | XP_016857774.1 | ||
XM_017002286.1 | 6340 | Intron | XP_016857775.1 | ||
XM_017002287.1 | 6340 | Intron | XP_016857776.1 | ||
XM_017002288.1 | 6340 | Intron | XP_016857777.1 |