Product Details

SNP ID
rs3825015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64823914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTCTACCTCCTTCCAATTCCTA[C/T]CACCTCCTCCTCCACTTCCTTCCTC
Phenotype
MIM: 613991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017525.2 5364 Intron NP_059995.2
XM_011545155.2 5364 UTR 3 XP_011543457.1
XM_011545156.2 5364 UTR 3 XP_011543458.1
XM_011545157.2 5364 UTR 3 XP_011543459.1
XM_011545158.2 5364 UTR 3 XP_011543460.1
XM_011545159.2 5364 Intron XP_011543461.1
XM_011545160.2 5364 UTR 3 XP_011543462.1
XM_011545161.2 5364 Intron XP_011543463.1
XM_017017996.1 5364 UTR 3 XP_016873485.1
XM_017017997.1 5364 Intron XP_016873486.1
XM_017017998.1 5364 Intron XP_016873487.1
XM_017017999.1 5364 UTR 3 XP_016873488.1

View Full Product Details