Product Details

SNP ID
rs12787211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64823940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCCTCCTCCACTTCCTTCCTC[A/C]TATCCTGTCTCTCACCTCCTTCGCC
Phenotype
MIM: 613991
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017525.2 5338 Intron NP_059995.2
XM_011545155.2 5338 UTR 3 XP_011543457.1
XM_011545156.2 5338 UTR 3 XP_011543458.1
XM_011545157.2 5338 UTR 3 XP_011543459.1
XM_011545158.2 5338 UTR 3 XP_011543460.1
XM_011545159.2 5338 Intron XP_011543461.1
XM_011545160.2 5338 UTR 3 XP_011543462.1
XM_011545161.2 5338 Intron XP_011543463.1
XM_017017996.1 5338 UTR 3 XP_016873485.1
XM_017017997.1 5338 Intron XP_016873486.1
XM_017017998.1 5338 Intron XP_016873487.1
XM_017017999.1 5338 UTR 3 XP_016873488.1

View Full Product Details