Product Details

SNP ID
rs11234042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71528040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCACCCCTCACCCGCTAGCCTTG[A/C]CTTTGCTTGTGTATTCAGAGGCCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-7 PubMed Links

Gene Details

Gene
KRTAP5-7
Gene Name
keratin associated protein 5-7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012503.1 Intron NP_001012521.1
Gene
KRTAP5-8
Gene Name
keratin associated protein 5-8
There are no transcripts associated with this gene.

View Full Product Details