Product Details
- SNP ID
-
hCV32066141
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:34957230 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCAGTTCCACTGAGCAACAGGAG[A/C]TAAGGGCTTTAAGCAGGGGAAGGAC
- Phenotype
-
MIM: 601002
MIM: 609928
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GSS
PubMed Links
Gene Details
- Gene
- GSS
- Gene Name
- glutathione synthetase
There are no transcripts associated with this gene.
- Gene
- MYH7B
- Gene Name
- myosin heavy chain 7B
There are no transcripts associated with this gene.
View Full Product Details