Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001207076.2 | 157 | Intron | NP_001194005.1 | ||
NM_001304367.1 | 157 | Intron | NP_001291296.1 | ||
NM_080834.3 | 157 | Intron | NP_543024.2 | ||
XM_005260295.2 | 157 | Intron | XP_005260352.1 | ||
XM_011528590.2 | 157 | Intron | XP_011526892.1 | ||
XM_011528592.1 | 157 | Intron | XP_011526894.1 | ||
XM_011528593.2 | 157 | Intron | XP_011526895.1 | ||
XM_011528598.1 | 157 | Intron | XP_011526900.1 | ||
XM_017027683.1 | 157 | Intron | XP_016883172.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_016558.3 | 157 | UTR 5 | NP_057642.1 | ||
NM_033630.2 | 157 | Intron | NP_361012.2 |