Product Details
- SNP ID
-
rs3812852
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:73057735 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATGAATAGGCTGTGGTATATGTAA[A/G]ACTGTCTAATGGTATATTGTGTGTC
- Phenotype
-
MIM: 602903
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLF5
PubMed Links
Gene Details
- Gene
- KLF5
- Gene Name
- Kruppel like factor 5
View Full Product Details