Product Details

SNP ID
rs10906249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12898455 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGGGGCGCACGGGCCCCCGGGC[A/G]TTGATGTGCGGCAGCGCGCCCGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC3 PubMed Links

Gene Details

Gene
CCDC3
Gene Name
coiled-coil domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282658.1 908 Silent Mutation NP_001269587.1
NM_031455.3 908 Silent Mutation NP_113643.1

View Full Product Details