Product Details

SNP ID
rs4403737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1069158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAACTTCCTGAGTATACTGAGAG[C/T]CATTGAATTGTGCACTTTAAATAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR37 PubMed Links

Gene Details

Gene
WDR37
Gene Name
WD repeat domain 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014023.3 Intron NP_054742.2

View Full Product Details