Product Details
- SNP ID
-
hCV32121469
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:1235942 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGACCACGTGCAGCGGCGAGCCCAT[G/T]GACCCGGAGGAGATCGCGCGCAAGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UNCX
PubMed Links
Gene Details
- Gene
- UNCX
- Gene Name
- UNC homeobox
There are no transcripts associated with this gene.
View Full Product Details