Product Details

SNP ID
rs10985830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122944590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCAGGCTGGAGTGTGGCGGCAC[C/G]ATATCAGCTCACTGCATCCTCCCAG
Phenotype
MIM: 615882
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RABGAP1 PubMed Links

Gene Details

Gene
RABGAP1
Gene Name
RAB GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012197.3 Intron NP_036329.3
XM_011518440.2 Intron XP_011516742.1
XM_011518441.2 Intron XP_011516743.1
XM_011518442.2 Intron XP_011516744.1
XM_011518444.2 Intron XP_011516746.1
XM_017014567.1 Intron XP_016870056.1
XM_017014568.1 Intron XP_016870057.1
XM_017014569.1 Intron XP_016870058.1
XM_017014570.1 Intron XP_016870059.1

View Full Product Details