Product Details

SNP ID
rs10818638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122138097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAAAGCCTTTAAGTCAAACCAC[C/T]GTCATATGACATAATACCTTAGTTT
Phenotype
MIM: 603359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA8 PubMed Links

Gene Details

Gene
NDUFA8
Gene Name
NADH:ubiquinone oxidoreductase subunit A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318195.1 Intron NP_001305124.1
NM_014222.2 Intron NP_055037.1

View Full Product Details