Product Details

SNP ID
rs13302571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122429725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGTAGTGAGCATTACATATGG[G/T]TTCCTTTGTATTTATCCTCCTTGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR1J2 PubMed Links

Gene Details

Gene
OR1J2
Gene Name
olfactory receptor family 1 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054107.1 Intron NP_473448.1
XM_017014623.1 Intron XP_016870112.1

View Full Product Details