Product Details
- SNP ID
-
rs4766004
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:2900373 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCACTTGGATTGGAAATGTTGCCT[A/G]CAGTAACCTGTATAAACATCAATAT
- Phenotype
-
MIM: 604730
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TULP3
PubMed Links
Gene Details
- Gene
- TULP3
- Gene Name
- tubby like protein 3
View Full Product Details