Product Details
- SNP ID
-
rs12056721
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:132712429 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAACAGCAGGTGATATAACCCC[C/T]GCAGATCGGATATTGGCTCTTCCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM71
PubMed Links
Gene Details
- Gene
- TMEM71
- Gene Name
- transmembrane protein 71
View Full Product Details