Product Details

SNP ID
rs13306087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63479049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATGAGCAGGATCTACTCCACC[A/G]CCAAGGTCTGCCTCCCCAACAAGAC
Phenotype
MIM: 106180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACE PubMed Links

Gene Details

Gene
ACE
Gene Name
angiotensin I converting enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000789.3 494 Missense Mutation ACC,GCC T154A NP_000780.1
NM_001178057.1 494 Intron NP_001171528.1
NM_152830.2 494 Intron NP_690043.1
XM_006721737.3 494 Intron XP_006721800.2

View Full Product Details