Product Details

SNP ID
rs3744086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57257290 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTAAATAGCAAATCCTTTCT[C/G]AGCCCTCTATGCGACCTCTGTTGCC
Phenotype
MIM: 607897
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MSI2 PubMed Links

Gene Details

Gene
MSI2
Gene Name
musashi RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322250.1 Intron NP_001309179.1
NM_001322251.1 Intron NP_001309180.1
NM_138962.3 Intron NP_620412.1
NM_170721.1 Intron NP_733839.1
XM_005257014.3 Intron XP_005257071.1
XM_005257015.3 Intron XP_005257072.1
XM_011524286.1 Intron XP_011522588.1
XM_011524287.2 Intron XP_011522589.1
XM_017024147.1 Intron XP_016879636.1
XM_017024148.1 Intron XP_016879637.1
XM_017024149.1 Intron XP_016879638.1

View Full Product Details