Product Details

SNP ID
rs7132172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13197311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTCAGGACTTGAACTGTTACTC[C/T]GCTCGCTGGTTGTTGGTTGGGATTT
Phenotype
MIM: 602333
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMP1 PubMed Links

Gene Details

Gene
EMP1
Gene Name
epithelial membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001423.2 Intron NP_001414.1

View Full Product Details