Product Details

SNP ID
rs12007581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85279153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGTTATGTTCTACAGTGTCTGC[A/C]ATCAAACTTTAAACAATCAGGTCCC
Phenotype
MIM: 300603 MIM: 314990
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POF1B PubMed Links

Gene Details

Gene
POF1B
Gene Name
premature ovarian failure, 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307940.1 2097 Intron NP_001294869.1
NM_024921.3 2097 UTR 3 NP_079197.3
XM_005262203.3 2097 UTR 3 XP_005262260.1
Gene
ZNF711
Gene Name
zinc finger protein 711
There are no transcripts associated with this gene.

View Full Product Details