Product Details
- SNP ID
-
rs12946339
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:16032244 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGACCTGCTACTAAAAATTAAACC[A/C]CAAAAACTAGAGATCCCTCTCCTGC
- Phenotype
-
MIM: 600849
MIM: 613814
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NCOR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112840784] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCOR1
- Gene Name
- nuclear receptor corepressor 1
- Gene
- TTC19
- Gene Name
- tetratricopeptide repeat domain 19
View Full Product Details