Product Details

SNP ID
rs6936410
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133091811 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAAGAGATCAATATCAGAATGT[A/T]TAAGAAACTTCTGCAAATTAAAATA
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
LINC00326 PubMed Links
Additional Information
For this assay, SNP(s) [rs147825366] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LINC00326
Gene Name
long intergenic non-protein coding RNA 326
There are no transcripts associated with this gene.

View Full Product Details