Product Details

SNP ID
rs11758072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75295035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAAAAAGATCCTGACATTTTA[A/G]GTCTGTTTTTATATTCAAGTTTTGA
Phenotype
MIM: 607307
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FILIP1 PubMed Links

Gene Details

Gene
FILIP1
Gene Name
filamin A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289987.1 4968 Intron NP_001276916.1
NM_001300866.1 4968 Intron NP_001287795.1
NM_015687.3 4968 Intron NP_056502.1
XM_005248713.3 4968 UTR 3 XP_005248770.1
XM_005248715.4 4968 UTR 3 XP_005248772.1
XM_011535756.2 4968 Intron XP_011534058.1
Gene
LOC100506804
Gene Name
uncharacterized LOC100506804
There are no transcripts associated with this gene.

View Full Product Details