Product Details

SNP ID
rs11619412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:109758676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACAGTTATCAAAACGTTAATTT[C/T]TAAGCATTAGCATTATGCAAGAAAA
Phenotype
MIM: 600797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRS2 PubMed Links

Gene Details

Gene
IRS2
Gene Name
insulin receptor substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003749.2 Intron NP_003740.2

View Full Product Details