Product Details

SNP ID
rs12873468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:108499074 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCCTGCCCTGGATGTGCTATTCA[G/T]TAAGGATTCAGTCAGTGCTTTCAGA
Phenotype
MIM: 615479
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYO16 PubMed Links

Gene Details

Gene
MYO16
Gene Name
myosin XVI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198950.1 Intron NP_001185879.1
NM_015011.1 Intron NP_055826.1
XM_011521062.1 Intron XP_011519364.1

View Full Product Details