Product Details

SNP ID
rs11674327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:173082225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCACTATGCTGCCCAAGTTGGT[C/T]TTAAATGCCTGACCTCAAGCGATAC
Phenotype
MIM: 609479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZAK PubMed Links

Gene Details

Gene
ZAK
Gene Name
sterile alpha motif and leucine zipper containing kinase AZK
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016653.2 Intron NP_057737.2
NM_133646.2 Intron NP_598407.1
XM_005246640.2 Intron XP_005246697.1
XM_017004323.1 Intron XP_016859812.1
XM_017004324.1 Intron XP_016859813.1

View Full Product Details