Product Details
- SNP ID
-
hCV32294048
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:77799180 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGTTTTATTTTTTGTGATGTCTT[G/T]GTCTGGCTTTGATATTAACTGTCCC
- Phenotype
-
MIM: 616773
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIGA1
PubMed Links
Gene Details
- Gene
- MIGA1
- Gene Name
- mitoguardin 1
View Full Product Details