Product Details

SNP ID
rs4253074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49515271 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAATTGGAACACTAGGCAAAACC[A/T]CTGCTACACTGTACATAATGTATAA
Phenotype
MIM: 609413
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ERCC6 PubMed Links

Gene Details

Gene
ERCC6
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000124.3 2100 Intron NP_000115.1
NM_001277058.1 2100 UTR 3 NP_001263987.1
NM_001277059.1 2100 UTR 3 NP_001263988.1
Gene
PGBD3
Gene Name
piggyBac transposable element derived 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170753.3 2100 UTR 3 NP_736609.2

View Full Product Details