Product Details

SNP ID
rs1052530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50378184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGAAGAGGAACCAGAGCGCAAG[A/C]GAAAGAAGGGCCCAGCCCCGAAGAT
Phenotype
MIM: 605631 MIM: 600380 MIM: 174761
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAPSA PubMed Links

Gene Details

Gene
NAPSA
Gene Name
napsin A aspartic peptidase
There are no transcripts associated with this gene.

Gene
NR1H2
Gene Name
nuclear receptor subfamily 1 group H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256647.1 498 Intron NP_001243576.1
NM_007121.5 498 Silent Mutation AGA,CGA R73R NP_009052.3
Gene
POLD1
Gene Name
DAN polymerase delta 1, catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details