Product Details
- SNP ID
-
rs4754261
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:107797396 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTGAGACAGAGAGAGAAGAAAAAG[A/C]AAGCAAGCTAGCTTGGCACAGTGGT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC35F2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs555493730] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC35F2
- Gene Name
- solute carrier family 35 member F2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017515.4 |
|
Intron |
|
|
NP_059985.2 |
View Full Product Details