Product Details
- SNP ID
-
rs4886795
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:76349877 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGCAGAGAGAGGCAGCTTCTGG[C/G]ACTGTGTCTTCCAGACAATGTAACA
- Phenotype
-
MIM: 609481
MIM: 611611
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ISL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs574002118] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ISL2
- Gene Name
- ISL LIM homeobox 2
There are no transcripts associated with this gene.
- Gene
- SCAPER
- Gene Name
- S-phase cyclin A associated protein in the ER
View Full Product Details