Product Details

SNP ID
rs4147551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99414750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTAAAAATGGTAAAAATAATA[C/T]CTGGCTTACAGGGATCTGAATAATG
Phenotype
MIM: 600086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADH7 PubMed Links

Gene Details

Gene
ADH7
Gene Name
alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000673.4 Intron NP_000664.2
NM_001166504.1 Intron NP_001159976.1

View Full Product Details