Product Details

SNP ID
rs3734929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130070734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGGTAGCGGGGCCGGCGGGGGC[A/C]GTGGGGGCAGCGGGGGTCGGGGGAC
Phenotype
MIM: 615152
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC10 PubMed Links

Gene Details

Gene
KLHDC10
Gene Name
kelch domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014997.3 202 Missense Mutation AGT,CGT S31R NP_055812.1
XM_005250233.4 202 Missense Mutation AGT,CGT S31R XP_005250290.1

View Full Product Details