Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318751.1 | 3346 | UTR 3 | NP_001305680.1 | ||
NM_001318752.1 | 3346 | UTR 3 | NP_001305681.1 | ||
NM_001318753.1 | 3346 | UTR 3 | NP_001305682.1 | ||
NM_001318754.1 | 3346 | UTR 3 | NP_001305683.1 | ||
NM_030820.3 | 3346 | UTR 3 | NP_110447.2 | ||
XM_006715223.1 | 3346 | UTR 3 | XP_006715286.1 | ||
XM_011514924.2 | 3346 | UTR 3 | XP_011513226.1 | ||
XM_011514925.2 | 3346 | UTR 3 | XP_011513227.1 | ||
XM_011514926.1 | 3346 | UTR 3 | XP_011513228.1 | ||
XM_011514927.1 | 3346 | UTR 3 | XP_011513229.1 |