Product Details

SNP ID
rs1051744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3082769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCGCGGGCGCGGCGGTGAAAG[A/C]CCTCGCGGCACTCGGGCTCGGTCAC
Phenotype
MIM: 192340
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AVP PubMed Links

Gene Details

Gene
AVP
Gene Name
arginine vasopressin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000490.4 406 Missense Mutation GGC,GTC G119V NP_000481.2
XM_011529267.1 406 Missense Mutation GGC,GTC G119V XP_011527569.1

View Full Product Details