Product Details

SNP ID
rs1136806
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:21217283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTATTTTATTTCCATTACAATAG[G/T]GCCACGCCAAAACTACAGTAGCTCA
Phenotype
MIM: 600423
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ECE1 PubMed Links

Gene Details

Gene
ECE1
Gene Name
endothelin converting enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113347.1 5205 UTR 3 NP_001106818.1
NM_001113348.1 5205 UTR 3 NP_001106819.1
NM_001113349.1 5205 UTR 3 NP_001106820.1
NM_001397.2 5205 UTR 3 NP_001388.1
XM_006710398.2 5205 UTR 3 XP_006710461.1
XM_011540872.2 5205 UTR 3 XP_011539174.1
XM_011540873.2 5205 UTR 3 XP_011539175.1
XM_017000511.1 5205 UTR 3 XP_016856000.1

View Full Product Details