Product Details

SNP ID
rs3212852
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50826873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCGCATCCAGCTCCATCCTTTCA[C/T]GCACCTTGTGACCGGGGGCTCCCAG
Phenotype
MIM: 147910 MIM: 610601
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KLK1 PubMed Links

Gene Details

Gene
KLK1
Gene Name
kallikrein 1
There are no transcripts associated with this gene.

Gene
KLK15
Gene Name
kallikrein related peptidase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277081.1 Intron NP_001264010.1
NM_001277082.1 Intron NP_001264011.1
NM_017509.3 Intron NP_059979.2
XM_006723265.3 Intron XP_006723328.1
XM_011527085.2 Intron XP_011525387.1
XM_011527087.2 Intron XP_011525389.1
XM_011527088.2 Intron XP_011525390.1
XM_011527089.2 Intron XP_011525391.1
XM_017026943.1 Intron XP_016882432.1
Gene
LOC105372441
Gene Name
uncharacterized LOC105372441
There are no transcripts associated with this gene.

Gene
MGC45922
Gene Name
uncharacterized LOC284365
There are no transcripts associated with this gene.

View Full Product Details