Product Details

SNP ID
rs3842725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2163956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACCACAGTTTCTTTTATTGTGA[C/T]GGTGATTGGGGCAGCAGACAGTGTC
Phenotype
MIM: 176730 MIM: 191290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INS PubMed Links

Gene Details

Gene
INS
Gene Name
insulin
There are no transcripts associated with this gene.

Gene
INS-IGF2
Gene Name
INS-IGF2 readthrough
There are no transcripts associated with this gene.

Gene
MIR4686
Gene Name
microRNA 4686
There are no transcripts associated with this gene.

Gene
TH
Gene Name
tyrosine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000360.3 4078 UTR 3 NP_000351.2
NM_199292.2 4078 UTR 3 NP_954986.2
NM_199293.2 4078 UTR 3 NP_954987.2
XM_011520335.2 4078 UTR 3 XP_011518637.1

View Full Product Details